Learn More
Medchemexpress LLC 1-cyclopropyl-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carbohydrazide | 112811-72-0 | MFCD05864420 | 99.6% | 295.24 g/mol | C14H11F2NO4 | 100 G
SDP
Catalog No. 5000300029
Encompass_Preferred
Antibacterial agent 265 (CAS 112811-72-0) is a small-molecule antibacterial research reagent active against Staphylococcus aureus, Micrococcus luteus, Bacillus subtilis, Escherichia coli, Pseudomonas aeruginosa, and Flavobacterium devorans. It is supplied as a white to off-white solid with high purity for laboratory research use.
  • High purity (99.59%).
  • Broad-spectrum antibacterial activity against Gram-positive and Gram-negative bacteria.
  • Solid form for accurate weighing and long-term storage.
  • Known molecular weight and formula for analytical and formulation work.
  • Stable when stored under recommended conditions.
Catalog No. 50-003-00029 Supplier Medchemexpress LLC Supplier No. HYW018629100G
Antibacterial agent 265 (CAS 112811-72-0) is a small-molecule antibacterial research reagent active against Staphylococcus aureus, Micrococcus luteus, Bacillus subtilis, Escherichia coli, Pseudomonas aeruginosa, and Flavobacterium devorans. It is supplied as a white to off-white solid with high purity for laboratory research use.
  • High purity (99.59%).
  • Broad-spectrum antibacterial activity against Gram-positive and Gram-negative bacteria.
  • Solid form for accurate weighing and long-term storage.
  • Known molecular weight and formula for analytical and formulation work.
  • Stable when stored under recommended conditions.