Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Cyclopropylcarbonyl)piperazine, 97%, Thermo Scientific™

Catalog No. AC454510050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC454510050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC454510050 Supplier Thermo Scientific Chemicals Supplier No. 454510050

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 59878-57-8
Molecular Formula C8H15N2O
Molecular Weight (g/mol) 155.22
MDL Number MFCD06369645
InChI Key KIALFUYSJAAJSU-UHFFFAOYSA-O
Synonym 1-cyclopropylcarbonyl piperazine, cyclopropyl piperazin-1-yl methanone, 1-cyclopropanecarbonylpiperazine, cyclopropyl-piperazin-1-yl-methanone, 1-cyclopropanecarbonyl piperazine, piperazine, 1-cyclopropylcarbonyl, piperazinocyclopropyl ketone, cyclopropyl piperazinyl ketone, 1-cyclopropylcarbonylpiperazine, 4-cyclopropanecarbonyl-piperazine
PubChem CID 2064235
IUPAC Name cyclopropyl(piperazin-1-yl)methanone
SMILES O=C(C1CC1)N1CC[NH2+]CC1

Specifications

Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 154.21
Percent Purity 97%
Chemical Name or Material 1-(Cyclopropylcarbonyl)piperazine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.