Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Cyclopropylmethyl)piperazine dihydrochloride, 97%, Thermo Scientific™

Catalog No. AAH6687003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6687003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6687003 Supplier Thermo Scientific Chemicals Supplier No. H6687003

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 373608-42-5
Molecular Formula C8H18Cl2N2
Molecular Weight (g/mol) 213.146
MDL Number MFCD08544420
InChI Key XOERBUIHFQNXBA-UHFFFAOYSA-N
Synonym 1-cyclopropylmethyl piperazine dihydrochloride, 1-cyclopropylmethylpiperazine dihydrochloride, 1-cyclopropylmethyl-piperazine dihydrochloride, c8h16n2.2clh, 1-cyclopropylmethyl piperazine 2hcl, 1-cyclopropylmethyl piperazine hydrochloride;piperazine, 1-cyclopropylmethyl piperazine-hydrogen chloride 1/2
PubChem CID 18403316
IUPAC Name 1-(cyclopropylmethyl)piperazine;dihydrochloride
SMILES C1CC1CN2CCNCC2.Cl.Cl

Specifications

Quantity 1 g
Beilstein 6420036
Formula Weight 213.15
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-(Cyclopropylmethyl)piperazine dihydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.