Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-deazaadenosine | 14432-09-8 | MFCD22683837 | 98.9% | 266.25 | C11H14N4O4 | 1 MG
SDP

Catalog No. 5000245591
Encompass_Preferred

1-Deazaadenosine is a nucleoside analogue used as a biochemical research reagent that potently inhibits adenosine deaminase (ADA) and is used in in vitro studies of ADA-related pathways and lymphoproliferative disorders.

  • Potent adenosine deaminase (ADA) inhibitor with Ki = 0.66 μM.
  • High purity: 98.9%.
  • Molecular weight: 266.25 g/mol.
  • Chemical formula: C11H14N4O4.
  • White to off-white solid suitable for laboratory use.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-45591 Supplier Medchemexpress LLC Supplier No. HY1031591MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1-Deazaadenosine is a nucleoside analogue used as a biochemical research reagent that potently inhibits adenosine deaminase (ADA) and is used in in vitro studies of ADA-related pathways and lymphoproliferative disorders.

  • Potent adenosine deaminase (ADA) inhibitor with Ki = 0.66 μM.
  • High purity: 98.9%.
  • Molecular weight: 266.25 g/mol.
  • Chemical formula: C11H14N4O4.
  • White to off-white solid suitable for laboratory use.
  • Recommended storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); in solvent at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.