Learn More
Medchemexpress LLC 1-Decanaminium, N,N-dimethyl-N-(3-sulfopropyl)-, inner salt | 15163-36-7 | 98.0% | 307.49 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY126377
Encompass_Preferred
Sulfobetaine 10 is a zwitterionic surfactant commonly used in biochemistry and biophysics. It is utilized to dissolve membrane proteins and other hydrophobic molecules in aqueous solutions. Its structure contains both positively and negatively charged groups, which allows it to maintain a neutral charge in solution and reduces its tendency to denature proteins. It is considered a mild, non-denaturing surfactant and is widely used in protein research. For research use only. We do not sell to patients.
  • Zwitterionic surfactant
  • Commonly used in biochemistry and biophysics
  • Dissolves membrane proteins and other hydrophobic molecules in aqueous solutions
  • Maintains a neutral charge in solution
  • Reduces tendency to denature proteins
  • Considered a mild, non-denaturing surfactant
  • Widely used in protein research
Catalog No. 50-004-39553