Learn More
Medchemexpress LLC 1-decanol | 112-30-1 | MFCD00004747 | 98.9% | 158.28 g/mol | C10H22O | 1kg
SDP
Catalog No. 5000267772
Encompass_Preferred
1-Decanol is used in the production of lubricants surfactants plasticizers and solvents It has also been used to study the thermal properties of polymeric monolithic stationary phases and to enhance homopolyglycine acceptor function Used in cosmetics daily flavors and food flavors
Catalog No. 50-002-67772 Supplier Medchemexpress LLC Supplier No. HYY10041KG
1-Decanol is used in the production of lubricants surfactants plasticizers and solvents It has also been used to study the thermal properties of polymeric monolithic stationary phases and to enhance homopolyglycine acceptor function Used in cosmetics daily flavors and food flavors