Learn More
Medchemexpress LLC 1-decanol | 112-30-1 | MFCD00004747 | 98.9% | 158.28 g/mol | C10H22O | 500g
SDP
Catalog No. 5000269633
Encompass_Preferred
1-Decanol is used in the production of lubricants surfactants plasticizers and solvents It has also been used to study the thermal properties of polymeric monolithic stationary phases and to enhance homopolyglycine acceptor function Used in cosmetics daily flavors and food flavors
Catalog No. 50-002-69633 Supplier Medchemexpress LLC Supplier No. HYY1004500G
1-Decanol is used in the production of lubricants surfactants plasticizers and solvents It has also been used to study the thermal properties of polymeric monolithic stationary phases and to enhance homopolyglycine acceptor function Used in cosmetics daily flavors and food flavors