Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Decyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide 98.0+%, TCI America™
SDP

Catalog No. D43515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43515G 5 g
D435125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43515G Supplier TCI America Supplier No. D43515G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 433337-23-6
Molecular Formula C16H27F6N3O4S2
Molecular Weight (g/mol) 503.519
MDL Number MFCD12022376
InChI Key ZYVGZWFCGPUVSH-UHFFFAOYSA-N
PubChem CID 12040509
IUPAC Name bis(trifluoromethylsulfonyl)azanide;1-decyl-3-methylimidazol-3-ium
SMILES CCCCCCCCCCN1C=C[N+](=C1)C.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F

Specifications

Melting Point -19°C
Color Yellow
Quantity 5 g
Formula Weight 503.52
Percent Purity ≥98.0% (HPLC,T)
Physical Form Liquid
Chemical Name or Material 1-Decyl-3-methylimidazolium Bis(trifluoromethanesulfonyl)imide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.