Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Decyl-3-methylimidazolium Chloride 96.0+%, TCI America™
SDP

Catalog No. D53515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53515G 5 g
D535125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53515G Supplier TCI America Supplier No. D53515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 171058-18-7
Molecular Formula C14H27ClN2
Molecular Weight (g/mol) 258.834
MDL Number MFCD03427605
InChI Key HTZVLLVRJHAJJF-UHFFFAOYSA-M
PubChem CID 2734225
ChEBI CHEBI:61348
IUPAC Name 1-decyl-3-methylimidazol-3-ium;chloride
SMILES CCCCCCCCCCN1C=C[N+](=C1)C.[Cl-]

Specifications

Quantity 5 g
Formula Weight 258.83
Percent Purity ≥96.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 1-Decyl-3-methylimidazolium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.