Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Decyl-3-Methylimidazolium Tetrafluoroborate

Catalog No. 69070850G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-070-850G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 69-070-850G Supplier Sigma Aldrich Supplier No. 69070850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Decyl-3-methylimidazolium tetrafluoroborate is an ionic liquid, which can be used as:A clathrate hydrate crystal inhibitor in drilling fluid.A microextraction solvent in the determination of synthetic dyes in foods and cosmetics.A substrate in the host-guest inclusion complexation studies with +a-cyclodextrin.

Specifications

CAS 6212-33-5
Molecular Formula C8H8ClNO2
Linear Formula C8H8ClNO2
Quantity 25 g
MDL Number MFCD00049328
Synonym [C10MIM][BF4]
Molecular Weight (g/mol) 185.61 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.