Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-deoxy-1-morpholino-D-fructose | 6291-16-3 | ≥97.0% | 249.26 g/mol | C10H19NO6 | 250 MG
SDP

Catalog No. 5000209444
Encompass_Preferred

1-Deoxy-1-morpholino-D-fructose is a fructose-derived biochemical reagent used in sugar science research and biochemical assays. It is a solid carbohydrate derivative with chemical formula C10H19NO6 and molecular weight 249.26 g/mol, typically supplied at ≥97.0% purity.

  • Suitable for biochemical assays and sugar chemistry research.
  • High purity (≥97.0%) reduces analytical interference.
  • Available in 100 mg, 250 mg, 500 mg, 1 g, and larger pack sizes.
  • Powder stability: -20°C (3 years) or 4°C (2 years); in solvent stability is shorter.
  • Molecular formula C10H19NO6 and molecular weight 249.26 g/mol.

Catalog No. 50-002-09444 Supplier Medchemexpress LLC Supplier No. 250MGHYW047388
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Deoxy-1-morpholino-D-fructose is a fructose-derived biochemical reagent used in sugar science research and biochemical assays. It is a solid carbohydrate derivative with chemical formula C10H19NO6 and molecular weight 249.26 g/mol, typically supplied at ≥97.0% purity.

  • Suitable for biochemical assays and sugar chemistry research.
  • High purity (≥97.0%) reduces analytical interference.
  • Available in 100 mg, 250 mg, 500 mg, 1 g, and larger pack sizes.
  • Powder stability: -20°C (3 years) or 4°C (2 years); in solvent stability is shorter.
  • Molecular formula C10H19NO6 and molecular weight 249.26 g/mol.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.