Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-deoxy-1-morpholino-D-fructose | 6291-16-3 | MFCD00166980 | ≥97.0% | 249.26 g/mol | C10H19NO6 | 500 MG
SDP

Catalog No. 5000210077
Encompass_Preferred

1-Deoxy-1-morpholino-D-fructose is a synthetic D-fructose derivative used as a reagent in carbohydrate and sugar chemistry research. It is supplied as a white to off-white solid powder with high purity and is provided in multiple pre-weighed laboratory sizes.

  • Used in sugar science and carbohydrate research.
  • White to off-white solid powder.
  • Purity ≥97.0%.
  • Molecular weight 249.26 g/mol and formula C10H19NO6.
  • Available in 100 mg, 250 mg, 500 mg, and 1 g sizes.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-10077 Supplier Medchemexpress LLC Supplier No. HYW047388500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Deoxy-1-morpholino-D-fructose is a synthetic D-fructose derivative used as a reagent in carbohydrate and sugar chemistry research. It is supplied as a white to off-white solid powder with high purity and is provided in multiple pre-weighed laboratory sizes.

  • Used in sugar science and carbohydrate research.
  • White to off-white solid powder.
  • Purity ≥97.0%.
  • Molecular weight 249.26 g/mol and formula C10H19NO6.
  • Available in 100 mg, 250 mg, 500 mg, and 1 g sizes.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.