Learn More
Medchemexpress LLC 1-Deoxy-1-morpholino-D-fructose | 6291-16-3 | MFCD00166980 | >98.0% | 249.26 g/mol | C10H19NO6 | 1 G
SDP
Catalog No. 5000210108
Encompass_Preferred
1-Deoxy-1-morpholino-D-fructose is a fructose derivative used as a biochemical research standard in carbohydrate chemistry and analytical assays. It is commonly used as a reference in fructosamine assays and for sugar science research; the compound has formula C10H19NO6 and molecular weight 249.26 g/mol and is supplied in small laboratory pack sizes.
  • Used as a reference standard in fructosamine and carbohydrate assays.
  • Applicable to sugar science and biochemical research applications.
  • Molecular formula C10H19NO6 and molecular weight 249.26 g/mol.
  • Provided in laboratory quantities suitable for analytical use.
Catalog No. 50-002-10108 Supplier Medchemexpress LLC Supplier No. HYW0473881G
1-Deoxy-1-morpholino-D-fructose is a fructose derivative used as a biochemical research standard in carbohydrate chemistry and analytical assays. It is commonly used as a reference in fructosamine assays and for sugar science research; the compound has formula C10H19NO6 and molecular weight 249.26 g/mol and is supplied in small laboratory pack sizes.
  • Used as a reference standard in fructosamine and carbohydrate assays.
  • Applicable to sugar science and biochemical research applications.
  • Molecular formula C10H19NO6 and molecular weight 249.26 g/mol.
  • Provided in laboratory quantities suitable for analytical use.