Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+)-1-Deoxymannojirimycin hydrochloride, Thermo Scientific Chemicals

Catalog No. AAJ61277LB0
Change view
Click to view available options
Quantity:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61277LB0 5 mg
AAJ61277MA 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ61277LB0 Supplier Thermo Scientific Chemicals Supplier No. J61277LB0
Add to Cart

Selective inhibitor of a-mannosidase

Selective inhibitor of a-mannosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73465-43-7
Molecular Formula C6H14ClNO4
Molecular Weight (g/mol) 199.631
MDL Number MFCD00083611
InChI Key ZJIHMALTJRDNQI-MVNLRXSJSA-N
Synonym DMJ; 1,5-Dideoxy-1,5-imino-D-mannitol hydrochloride
PubChem CID 11390018
IUPAC Name (2R,3R,4R,5R)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
SMILES C1C(C(C(C(N1)CO)O)O)O.Cl

Specifications

Type (+)-1-Deoxymannojirimycin Hydrochloride
Physical Form Powder
Color White
Quantity 5 mg
Formula Weight 199.6
Chemical Name or Material (+)-1-Deoxymannojirimycin hydrochloride
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.