Learn More
Cayman Chemical 1-DeoxymnojIrImycInhydroc 10mg
SDP
Catalog No. 502967292
Encompass_Preferred
A specific inhibitor of class I α-1,2-mannosidase (IC50 = 20 mM), a key enzyme for N-glycan processing in the endoplasmic reticulum and Golgi; inhibiting α-1,2-mannosidase with 1-Deoxymannojirimycin generates N-linked oligosaccharides with high mannose content, preventing proteasomal degradation of misfolded proteins
Catalog No. 50-296-7292 Supplier Cayman Chemical Supplier No. 1717810 MG
A specific inhibitor of class I α-1,2-mannosidase (IC50 = 20 mM), a key enzyme for N-glycan processing in the endoplasmic reticulum and Golgi; inhibiting α-1,2-mannosidase with 1-Deoxymannojirimycin generates N-linked oligosaccharides with high mannose content, preventing proteasomal degradation of misfolded proteins