Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Dicyanomethylene)-3-iminoisoindoline 98.0+%, TCI America™
SDP

Catalog No. D20801G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D20801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D20801G Supplier TCI America Supplier No. D20801G

Chemical Identifiers

CAS 43002-19-3
Molecular Formula C11H6N4
Molecular Weight (g/mol) 194.197
MDL Number MFCD00142610
InChI Key BDXWTIVCFJPDDL-UHFFFAOYSA-N
PubChem CID 606579
IUPAC Name 2-(3-aminoisoindol-1-ylidene)propanedinitrile
SMILES C1=CC=C2C(=C1)C(=C(C#N)C#N)N=C2N

Specifications

Color Yellow
Quantity 1 g
UN Number 3439
Formula Weight 194.20
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Dicyanomethylene)-3-iminoisoindoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.