Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(Dimethylamino)-2-nitroethylene | 1190-92-7 | 99.88% | 116.12 | 500 G
SDP

Catalog No. 5000401014
Encompass_Preferred

1-(Dimethylamino)-2-nitroethylene is a drug intermediate used for the synthesis of various active compounds. This product has a high purity of 99.88% and appears as a solid with a brown to dark brown color.

  • High purity of 99.88%
  • Appears as a solid
  • Color is brown to dark brown
  • Soluble in DMSO at ≥ 200 mg/mL
  • Molecular weight is 116.12
  • Chemical formula is C4H8N2O2
  • CAS number is 1190-92-7

Catalog No. 50-004-01014 Supplier Medchemexpress LLC Supplier No. HY20395500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(Dimethylamino)-2-nitroethylene is a drug intermediate used for the synthesis of various active compounds. This product has a high purity of 99.88% and appears as a solid with a brown to dark brown color.

  • High purity of 99.88%
  • Appears as a solid
  • Color is brown to dark brown
  • Soluble in DMSO at ≥ 200 mg/mL
  • Molecular weight is 116.12
  • Chemical formula is C4H8N2O2
  • CAS number is 1190-92-7

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.