Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(Dimethylamino)-2-nitroethylene | 1190-92-7 | 99.9% | 116.12 | 1 KG
SDP

Catalog No. 5000400982
Encompass_Preferred

1-(Dimethylamino)-2-nitroethylene is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.

  • Purity: 99.88%
  • CAS number: 1190-92-7
  • Molecular weight: 116.12
  • Formula: C4H8N2O2
  • Appearance: Solid, brown to dark brown
  • Ships at room temperature
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • Solvent storage: -80°C for 6 months; -20°C for 1 month
  • Soluble in DMSO (≥ 200 mg/mL)

Catalog No. 50-004-00982 Supplier Medchemexpress LLC Supplier No. HY203951KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(Dimethylamino)-2-nitroethylene is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.

  • Purity: 99.88%
  • CAS number: 1190-92-7
  • Molecular weight: 116.12
  • Formula: C4H8N2O2
  • Appearance: Solid, brown to dark brown
  • Ships at room temperature
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • Solvent storage: -80°C for 6 months; -20°C for 1 month
  • Soluble in DMSO (≥ 200 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.