Learn More
1-Dimethylamino-2-propanol, 99%
Description
1-Dimethylamino-2-propanol is one of the compounds used to identify the illegal manufacture of methadone.cv
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Dimethylamino-2-propanol is one of the compounds used to identify the illegal manufacture of methadone.cv
Solubility
Fully miscible in water.
Notes
Keep container tightly closed. Store in cool dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air. Store under dry inert gas.
Specifications
Specifications
| Melting Point | -85°C |
| Density | 0.85 |
| pH | 11 |
| Boiling Point | 123°C to 126°C |
| Flash Point | 35°C (95°F) |
| Odor | Amine-like |
| Refractive Index | 1.4195 |
| Quantity | 1000 g |
| UN Number | UN2734 |
| Sensitivity | Air Sensitive |
| Show More |
Safety and Handling
| Hazard Category | H226-H302-H314 |
| Hazard Statement | GHS H Statement H314-H226-H302 Causes severe skin burns and eye damage. Flammable liquid and vapour. Harmful if swallowed. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S. |
| EINECSNumber | 203-556-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.