Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dimethylamino-2-propanol, 99%

Catalog No. p-115306
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC115872500 250 g Glass bottle
AC115870010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC115872500 Supplier Thermo Scientific Chemicals Supplier No. 115872500
Only null left
Add to Cart

CAS: 108-16-7 | C5H13NO | 103.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-16-7
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.16
MDL Number MFCD00004532
InChI Key NCXUNZWLEYGQAH-UHFFFAOYSA-N
Synonym 1-dimethylamino-2-propanol, dimepranol, 1-dimethylamino propan-2-ol, n,n-dimethylisopropanolamine, dimethyl 2-hydroxypropyl amine, 2-propanol, 1-dimethylamino, dimethylisopropanolamine, dimepranol inn, n,n-dimethylamino-2-propanol
PubChem CID 37511
IUPAC Name 1-(dimethylamino)propan-2-ol
SMILES CC(CN(C)C)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -85.0°C
Color Colorless to Yellow
Density 0.8500g/mL
Boiling Point 126.0°C
Flash Point 26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4183 to 1.4203
Linear Formula (CH3)2NCH2CH(OH)CH3
Quantity 250 g
Specific Gravity 0.85
Solubility Information Solubility in water: miscible with water. Other solubilities: soluble in most organic solvents
Viscosity 4.9 mPa.s (50°C)
Formula Weight 103.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Dimethylamino-2-propanol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa
EINECSNumber 203-556-4
RTECSNumber UB3150000
TSCA TSCA
Recommended Storage Product may turn yellow on prolonged storage

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.