Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dimethylamino-but-1-en-3-one 99+%, Thermo Scientific™

Catalog No. AC408051000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC408051000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC408051000 Supplier Thermo Scientific Chemicals Supplier No. 408051000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2802-08-6
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
InChI Key QPWSKIGAQZAJKS-SNAWJCMRSA-N
Synonym e-4-dimethylamino but-3-en-2-one, trans-4-dimethylamino-3-buten-2-one, 4-dimethylamino but-3-en-2-one, 3e-4-dimethylamino but-3-en-2-one, 1-dimethylamino-but-1-en-3-one, 4-dimethylamino-3-buten-2-one, 3e-4-dimethylamino-3-buten-2-one, 3-buten-2-one, 4-dimethylamino-, 3e, 3-buten-2-one, 4-dimethylamino, 3-buten-2-one, 4-dimethylamino-, 3e-9ci
PubChem CID 5369152
IUPAC Name (E)-4-(dimethylamino)but-3-en-2-one
SMILES CC(=O)C=CN(C)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Brown to Brown-Red
Density 0.96
Infrared Spectrum Authentic
Refractive Index 1.5550 to 1.5580 (20°C, 589nm)
Linear Formula (CH3)2NCHCHC(O)CH3
Quantity 100 g
Formula Weight 113.16
Percent Purity ≥99.0%
Physical Form Liquid
Chemical Name or Material 1-Dimethylamino-but-1-en-3-one, 99+%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.