Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Diphenylmethyl)-3-hydroxyazetidine 98.0+%, TCI America™
SDP

Catalog No. D39075G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D39075G 5 g
D390725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D39075G Supplier TCI America Supplier No. D39075G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 18621-17-5
Molecular Formula C16H17NO
Molecular Weight (g/mol) 239.32
MDL Number MFCD00205109
InChI Key MMAJXKGUZYDTHV-UHFFFAOYSA-N
Synonym 1-benzhydrylazetan-3-ol, 1-diphenylmethyl-3-hydroxyazetidine, 1-diphenylmethyl azetidin-3-ol, 1-benzhydryl-3-azetidinol, 1-benzhydryl-3-hydroxyazetidine, 1-benzhydryl-azetidin-3-ol, n-benzhydrylazetidin-3-ol, 3-azetidinol, 1-diphenylmethyl, 1-diphenylmethyl-3-azetidinol
PubChem CID 330448
IUPAC Name 1-(diphenylmethyl)azetidin-3-ol
SMILES OC1CN(C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 5 g
Formula Weight 239.32
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(Diphenylmethyl)-3-hydroxyazetidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.