Learn More
Sigma Aldrich Fine Chemicals Biosciences 1-Dodecanol reagent grade100G
Catalog No. 502898558
Encompass
1-Dodecanol a fatty alcohol is a long chain aliphatic alcohol. Solid-liquid phase diagrams of its binary mixtures with various other fatty alcohols (1-tetradecanol 1-hexadecanol and 1-octadecanol) have been evaluated by employing high pressure microscopy and DSC (Differential scanning calorimetry). It undergoes oxidation in the presence of multiwall carbon nanotubes (MWCNTs) supported MoO3 (Mo/MWCNTs) and potassium containing Mo/MWCNTs (K/Mo/MWCNTs) catalysts to afford 1-dodecanal. Its oxidation in the presence of Ru and Pt based carbon catalysts has been reported.
Catalog No. 50-289-8558 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 126799100G
1-Dodecanol a fatty alcohol is a long chain aliphatic alcohol. Solid-liquid phase diagrams of its binary mixtures with various other fatty alcohols (1-tetradecanol 1-hexadecanol and 1-octadecanol) have been evaluated by employing high pressure microscopy and DSC (Differential scanning calorimetry). It undergoes oxidation in the presence of multiwall carbon nanotubes (MWCNTs) supported MoO3 (Mo/MWCNTs) and potassium containing Mo/MWCNTs (K/Mo/MWCNTs) catalysts to afford 1-dodecanal. Its oxidation in the presence of Ru and Pt based carbon catalysts has been reported.