Learn More
Medchemexpress LLC 1-dodecene | 112-41-4 | MFCD00008961 | 96.9% | 168.32 g/mol | C12H24 | 100 G
SDP
Catalog No. 5000300890
Encompass_Preferred
1-Dodecene is a linear alpha-olefin (C12H24) used as a reagent and chemical intermediate in organic synthesis and in the manufacture of surfactants and lubricants. It is supplied as a colorless to light yellow liquid with a GC-reported purity of 96.9% and a molecular weight of 168.32 g/mol.
  • Linear alpha-olefin reagent for organic synthesis and intermediates.
  • High GC purity (96.9%).
  • Colorless to light yellow liquid, easy to handle in standard laboratory conditions.
  • Molecular formula C12H24; molecular weight 168.32 g/mol.
  • Suitable for use in surfactant and lubricant production.
  • Certificate of analysis and safety data sheet available for quality and safety verification.
Catalog No. 50-003-00890 Supplier Medchemexpress LLC Supplier No. HYW017072100G
1-Dodecene is a linear alpha-olefin (C12H24) used as a reagent and chemical intermediate in organic synthesis and in the manufacture of surfactants and lubricants. It is supplied as a colorless to light yellow liquid with a GC-reported purity of 96.9% and a molecular weight of 168.32 g/mol.
  • Linear alpha-olefin reagent for organic synthesis and intermediates.
  • High GC purity (96.9%).
  • Colorless to light yellow liquid, easy to handle in standard laboratory conditions.
  • Molecular formula C12H24; molecular weight 168.32 g/mol.
  • Suitable for use in surfactant and lubricant production.
  • Certificate of analysis and safety data sheet available for quality and safety verification.