Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-dodecene | 112-41-4 | MFCD00008961 | 96.9% | 168.32 g/mol | C12H24 | 500 G
SDP

Catalog No. 5000300334
Encompass_Preferred

1-Dodecene (CAS 112-41-4) is a linear alpha-olefin supplied as a colorless to light yellow liquid. It serves as a chemical intermediate for organic synthesis and is commonly used in polymer production, lubricant formulation, and surfactant manufacturing.

  • Linear alpha-olefin C12H24 used as a chemical intermediate.
  • Colorless to light yellow liquid with density 0.7584 g/cm³.
  • Molecular weight 168.32 g/mol.
  • Purity 96.9% (as supplied).
  • Common applications include polymer synthesis, lubricant, and surfactant production.
  • Available in lab pack sizes including 100 g and 500 g, bulk quantities by quote.
  • Documentation available: COA and SDS.

Catalog No. 50-003-00334 Supplier Medchemexpress LLC Supplier No. HYW017072500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Dodecene (CAS 112-41-4) is a linear alpha-olefin supplied as a colorless to light yellow liquid. It serves as a chemical intermediate for organic synthesis and is commonly used in polymer production, lubricant formulation, and surfactant manufacturing.

  • Linear alpha-olefin C12H24 used as a chemical intermediate.
  • Colorless to light yellow liquid with density 0.7584 g/cm³.
  • Molecular weight 168.32 g/mol.
  • Purity 96.9% (as supplied).
  • Common applications include polymer synthesis, lubricant, and surfactant production.
  • Available in lab pack sizes including 100 g and 500 g, bulk quantities by quote.
  • Documentation available: COA and SDS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.