Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dodecylnaphthalene 97.0+%, TCI America™
SDP

Catalog No. D21451G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D21451G 1 g
D2145500MG 500 mg
D21455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D21451G Supplier TCI America Supplier No. D21451G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 38641-16-6
Molecular Formula C22H32
Molecular Weight (g/mol) 296.50
MDL Number MFCD00142676
InChI Key UVMGANPLBFFQIO-UHFFFAOYSA-N
PubChem CID 161973
IUPAC Name 1-dodecylnaphthalene
SMILES CCCCCCCCCCCCC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 30°C
Color White
Boiling Point 415°C
Quantity 1 g
Formula Weight 296.50
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Dodecylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.