Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Dodecyloxy-4-nitrobenzene 98.0+%, TCI America™
SDP

Catalog No. D202925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D202925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D202925G Supplier TCI America Supplier No. D202925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65039-18-1
Molecular Formula C18H29NO3
Molecular Weight (g/mol) 307.434
MDL Number MFCD00024678
InChI Key VZOAOBWVYOLSII-UHFFFAOYSA-N
Synonym Dodecyl 4-Nitrophenyl Ether, 4-Dodecyloxynitrobenzene, Lauryl 4-Nitrophenyl Ether, 1-Lauryloxy-4-nitrobenzene
PubChem CID 144094
IUPAC Name 1-dodecoxy-4-nitrobenzene
SMILES CCCCCCCCCCCCOC1=CC=C(C=C1)[N+](=O)[O-]

Specifications

Melting Point 51°C
Color White-Yellow
Boiling Point 220°C
Quantity 25 g
Formula Weight 307.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Dodecyloxy-4-nitrobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.