Learn More
Analytical Sales & Services 1 DRAM, 24-Position Parallel Synthesis Reaction Block with 18mm Spacing, Open Top
Catalog No. 5000806561
Encompass
 Gen II, 24 Position, 5-BOLT, OPEN TOP, Parallel Synthesis Reaction Block\n• For 15mm OD (1 Dram) Vials\n• 18mm Well Spacing\n• Includes Vial Rack, Open Top Cover, PFA Film, Rubber Mats and Screws (vials not included)\n\n 
Catalog No. 50-008-06561 Supplier Analytical Sales & Services Supplier No. 109240
 Gen II, 24 Position, 5-BOLT, OPEN TOP, Parallel Synthesis Reaction Block\n• For 15mm OD (1 Dram) Vials\n• 18mm Well Spacing\n• Includes Vial Rack, Open Top Cover, PFA Film, Rubber Mats and Screws (vials not included)\n\n 