Learn More
Analytical Sales & Services 1 DRAM, 24-Position TOP-DOWN Photoredox Reaction Block with 18mm Spacing, Gen II
Catalog No. 5000807266
Encompass
Para-Dox Gen II, 24 Position, TOP-DOWN, 5-BOLT, Photoredox Reaction Block\n• For 4mL (1 Dram), 15x45mm  Vials\n• 18mm Well Spacing\n• Includes Vial Rack, Top Cover (lid), Clear Top Sealing Mat, PFA Film, Mylar Bottom Film, Bottom Cover and Screws (vials not included)
Catalog No. 50-008-07266 Supplier Analytical Sales & Services Supplier No. 111240
Para-Dox Gen II, 24 Position, TOP-DOWN, 5-BOLT, Photoredox Reaction Block\n• For 4mL (1 Dram), 15x45mm  Vials\n• 18mm Well Spacing\n• Includes Vial Rack, Top Cover (lid), Clear Top Sealing Mat, PFA Film, Mylar Bottom Film, Bottom Cover and Screws (vials not included)