Learn More
Ace Glass, Inc. 1 dram poly-seal vial, cs/2304, sp/144
SDP
Catalog No. 50976407
Encompass
1 dram sample vials fabricated from borosilicate glass. Include poly-seal cone-lined closures, which are molded in a premium grade, low density polyethylene. Inverted cone liner provides excellent seal and torque retention. Pack/144.
Catalog No. 50-976-407 Supplier Ace Glass, Inc. Supplier No. 877910
1 dram sample vials fabricated from borosilicate glass. Include poly-seal cone-lined closures, which are molded in a premium grade, low density polyethylene. Inverted cone liner provides excellent seal and torque retention. Pack/144.