Learn More
Uline Spill Containment Workstation - 1 Drum | N/A | 1 EA
Catalog No. NC2211091
Encompass
Low-profile workstations protect floors from spills and leaks from one drum, Configure drum handling and dispensing areas, Tough high-density polyethylene construction, Resists chemicals rust and corrosion, Withstands extreme temperatures -20 to 120F, Non-skid easy-to-clean removable grates, Meets EPA SPCC and NPDES regulations
This item is not returnable. View return policy
Catalog No. NC2211091 Supplier Uline Supplier No. H4865
Low-profile workstations protect floors from spills and leaks from one drum, Configure drum handling and dispensing areas, Tough high-density polyethylene construction, Resists chemicals rust and corrosion, Withstands extreme temperatures -20 to 120F, Non-skid easy-to-clean removable grates, Meets EPA SPCC and NPDES regulations