Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-[(E)-(4-bromophenyl)methylideneamino]-3-(2,6-dimethylphenyl)urea | 415687-81-9 | MFCD28118904 | 99.8% | 346.22 g/mol | C16H16BrN3O | 5MG
SDP

Catalog No. 5000235452
Encompass_Preferred

EGA is a small-molecule inhibitor that selectively disrupts endosomal trafficking pathways exploited by toxins and viruses. It is intended for research use in cell-based assays to investigate intracellular trafficking, viral entry, and toxin uptake. Handle and store according to institutional safety procedures and the product safety data sheet.

  • Selective endosomal trafficking inhibitor used in cell-based assays.
  • CAS number 415687-81-9.
  • Chemical formula C16H16BrN3O.
  • Molecular weight 346.22 g/mol.
  • High purity: 99.8%.
  • Available in small-mass research pack sizes for assay preparation.

Catalog No. 50-002-35452 Supplier Medchemexpress LLC Supplier No. HY1251685MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

EGA is a small-molecule inhibitor that selectively disrupts endosomal trafficking pathways exploited by toxins and viruses. It is intended for research use in cell-based assays to investigate intracellular trafficking, viral entry, and toxin uptake. Handle and store according to institutional safety procedures and the product safety data sheet.

  • Selective endosomal trafficking inhibitor used in cell-based assays.
  • CAS number 415687-81-9.
  • Chemical formula C16H16BrN3O.
  • Molecular weight 346.22 g/mol.
  • High purity: 99.8%.
  • Available in small-mass research pack sizes for assay preparation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.