Learn More
Medchemexpress LLC 1-Eicosanol | 629-96-9 | MFCD00002938 | 298.55 | 500 MG
SDP
Catalog No. 5000448581
Encompass_Preferred
1-Eicosanol (Arachidyl alcohol) is a long-chain fatty alcohol and volatile organic compound. It can be isolated from natural sources and is applicable to research related to plant diseases such as tomato early blight and tomato gray mold.
  • Weakly inhibits mycelial growth of *Alternaria solani*
  • Very weakly inhibits mycelial growth of *Botrytis cinerea*
  • Volatile organic compound
  • Applicable for research in plant pathology
Catalog No. 50-004-48581 Supplier Medchemexpress LLC Supplier No. HYW004263500MG
1-Eicosanol (Arachidyl alcohol) is a long-chain fatty alcohol and volatile organic compound. It can be isolated from natural sources and is applicable to research related to plant diseases such as tomato early blight and tomato gray mold.
  • Weakly inhibits mycelial growth of *Alternaria solani*
  • Very weakly inhibits mycelial growth of *Botrytis cinerea*
  • Volatile organic compound
  • Applicable for research in plant pathology