Learn More
Medchemexpress LLC 1-Eicosanol-d41 | 349553-89-5 | 339.80 | 5 MG
SDP
Catalog No. 5000447473
Encompass_Preferred
1-Eicosanol-d41 (Arachidyl alcohol-d41) is the deuterium labeled 1-Eicosanol, a natural compound with antioxidant activity found in *Justicia insularis* leaves. Deuteration, which involves incorporating stable heavy isotopes into drug molecules, can affect their pharmacokinetic and metabolic profiles. This compound is suitable for use as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits antioxidant activity
  • Potential to influence pharmacokinetic and metabolic drug profiles
Catalog No. 50-004-47473 Supplier Medchemexpress LLC Supplier No. HYW004263S5MG
1-Eicosanol-d41 (Arachidyl alcohol-d41) is the deuterium labeled 1-Eicosanol, a natural compound with antioxidant activity found in *Justicia insularis* leaves. Deuteration, which involves incorporating stable heavy isotopes into drug molecules, can affect their pharmacokinetic and metabolic profiles. This compound is suitable for use as a tracer or as an internal standard for quantitative analysis.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Exhibits antioxidant activity
  • Potential to influence pharmacokinetic and metabolic drug profiles