Learn More
Medchemexpress LLC 1-Eicosanol-d41 (Arachidyl alcohol-d41) | 349553-89-5 | 339.80 | 10 MG
SDP
Catalog No. 5000445748
Encompass_Preferred
1-Eicosanol-d41 (Arachidyl alcohol-d41) is a deuterium-labeled version of 1-Eicosanol, which is a natural compound found in Justicia insularis leaves known for its antioxidant activity. Deuterium-labeled compounds are primarily used as tracers in the drug development process for quantitation and can potentially affect the pharmacokinetic and metabolic profiles of drugs.
  • Application as tracer
  • Internal standard for quantitative analysis
  • Antioxidant properties
Catalog No. 50-004-45748 Supplier Medchemexpress LLC Supplier No. HYW004263S10MG
1-Eicosanol-d41 (Arachidyl alcohol-d41) is a deuterium-labeled version of 1-Eicosanol, which is a natural compound found in Justicia insularis leaves known for its antioxidant activity. Deuterium-labeled compounds are primarily used as tracers in the drug development process for quantitation and can potentially affect the pharmacokinetic and metabolic profiles of drugs.
  • Application as tracer
  • Internal standard for quantitative analysis
  • Antioxidant properties