Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Eicosene, MP Biomedicals™

Catalog No. ICN15790950 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15790950 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15790950 Supplier MP Biomedicals, Inc Supplier No. 0215790950
Add to Cart
Add to Cart

Chemical Identifiers

CAS 9-3-3452
Molecular Formula C9H16
Molecular Weight (g/mol) 124.23
InChI Key OSSQSXOTMIGBCF-UHFFFAOYSA-N
Synonym 1-nonyne, nonyne, unii-760v9gp5a1, 3-acetylnaphthalene, pubchem13060, acmc-1aglr, 1-nonyne 6ci, 7ci, 8ci, 9ci
PubChem CID 18937
IUPAC Name non-1-yne
SMILES CCCCCCCC#C

Specifications

Color Colorless
Assay Percent Range 99.2%
Quantity 50 g
Formula Weight 280.5
Physical Form Liquid After Melting
Chemical Name or Material 1-Eicosene
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.