Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Eicosene 85.0+%, TCI America™
SDP

Catalog No. E000525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E000525ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E000525ML Supplier TCI America Supplier No. E000525ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7-1-3452
Molecular Formula C20H40
Molecular Weight (g/mol) 280.54
MDL Number MFCD00009013
InChI Key VAMFXQBUQXONLZ-UHFFFAOYSA-N
Synonym 1-eicosene, 1-icosene, eicosene, cetyl ethylene, .alpha.-eicosene, unii-6bhh7zd637, arachidene, arachydene, cosen, alpha-eicosene
PubChem CID 18936
IUPAC Name icos-1-ene
SMILES CCCCCCCCCCCCCCCCCCC=C

Specifications

Color White-Yellow
Boiling Point 151°C
Quantity 25 mL
Formula Weight 280.54
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Eicosene
EINECSNumber (2)-0027
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.