Learn More
Medchemexpress LLC 1'-epi Gemcitabine hydrochloride | 122111-05-1 | C9H12ClF2N3O4 | 5 MG
SDP
Catalog No. 5000432936
Encompass_Preferred
1'-epi Gemcitabine hydrochloride is an isomer of Gemcitabine hydrochloride and can be used as an experimental control. It is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent that inhibits DNA synthesis and repair, resulting in autophagy and apoptosis.
  • Isomer of Gemcitabine hydrochloride
  • Can be used as an experimental control
  • Pyrimidine nucleoside analog antimetabolite
  • Antineoplastic agent
  • Inhibits DNA synthesis and repair
  • Results in autophagy and apoptosis
  • For research use only
Catalog No. 50-004-32936 Supplier Medchemexpress LLC Supplier No. HYB0003A5MG
1'-epi Gemcitabine hydrochloride is an isomer of Gemcitabine hydrochloride and can be used as an experimental control. It is a pyrimidine nucleoside analog antimetabolite and an antineoplastic agent that inhibits DNA synthesis and repair, resulting in autophagy and apoptosis.
  • Isomer of Gemcitabine hydrochloride
  • Can be used as an experimental control
  • Pyrimidine nucleoside analog antimetabolite
  • Antineoplastic agent
  • Inhibits DNA synthesis and repair
  • Results in autophagy and apoptosis
  • For research use only