Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Ethoxy-2-propanol

Catalog No. 901255500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-125-5500MG 500 mg
90-125-5250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-125-5500MG Supplier Sigma Aldrich Supplier No. 901255500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Useful building block for synthesis
  • 1-ethoxy-2-propanol may be used in the preparation of 1-ethoxy-2-bromopropane using phosphorus tribromide
  • This product was previously listed as cds000221.reaction of 2-alkoxymethylthiiranes with lithium aluminum hydride and with secondary amines

Specifications

CAS 1569-02-4
Density 0.898 g/mL
Molecular Formula C5H12O2
Linear Formula C5H12O2
Refractive Index n/D 1.407
MDL Number MFCD00067050
Quantity 500 mg
Synonym 1-Ethoxypropan-2-ol
Molecular Weight (g/mol) 104.15
Percent Purity ≥95%
Biological Activity Central Nervous System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.