Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich 1-Ethyl-1-Methylpyrrolidinium Bromide
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
1-Ethyl-1-methylpyrrolidinium bromide is an ionic liquid that can be prepared by reacting N-methylpyrrolidine with bromoethane. it is conventionally used as a bromine-sequestering agent (BSA) in zinc/bromine redox flow batteries.1-Ethyl-1-methylpyrrolidinium bromide may be used in the preparation of 1-ethyl-1-methylpyrrolidiniu mLactate, a potential solvent for the separation of hexan-1-ene from hexane. It can also be used to prepare 1-ethyl-1-methylpyrrolidinium tribromide. It can be used as a co-catalyst during the cycloaddition of CO2 with different epoxides to form cyclic carbonates in the presence of a nona-vacant Keggin-type tricarbonyl rhenium derivative catalyst.
Specifications
Specifications
| CAS | 69227-51-6 |
| Molecular Formula | C7H16BrN |
| Linear Formula | C7H16BrN |
| MDL Number | MFCD03095384 |
| Quantity | 50 g |
| Molecular Weight (g/mol) | 194.11 g/mol |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?