Learn More
Medchemexpress LLC 1-ethyl-1-methylpyrrolidinium tetrafluoroborate | 117947-85-0 | MFCD08458505 | >98.0% | 201.01 | C7H16BF4N | 5 G
SDP
Catalog No. 5000283106
Encompass_Preferred
1-Ethyl-1-methylpyrrolidinium tetrafluoroborate is an ionic liquid supplied as a white to off-white solid used as a reagent in organic synthesis, electrochemistry, and laboratory research. It delivers a stable tetrafluoroborate counterion for low-volatility, thermally stable ionic media and is commonly used as an electrolyte or reaction medium in materials and synthesis workflows.
  • Ionic liquid with tetrafluoroborate anion.
  • Solid, white to off-white appearance.
  • Molecular formula C7H16BF4N; molecular weight 201.01.
  • High purity (>98.0%).
  • Suitable for organic synthesis and electrochemical applications.
  • Available in small laboratory pack sizes for research use.
Catalog No. 50-002-83106 Supplier Medchemexpress LLC Supplier No. HYW1277005G
1-Ethyl-1-methylpyrrolidinium tetrafluoroborate is an ionic liquid supplied as a white to off-white solid used as a reagent in organic synthesis, electrochemistry, and laboratory research. It delivers a stable tetrafluoroborate counterion for low-volatility, thermally stable ionic media and is commonly used as an electrolyte or reaction medium in materials and synthesis workflows.
  • Ionic liquid with tetrafluoroborate anion.
  • Solid, white to off-white appearance.
  • Molecular formula C7H16BF4N; molecular weight 201.01.
  • High purity (>98.0%).
  • Suitable for organic synthesis and electrochemical applications.
  • Available in small laboratory pack sizes for research use.