Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-ethyl-1H-pyrazole-5-carboxylic acid-d5 | 2735654-39-2 | 99.9% | 145.17 | C6H3D5N2O2 | 50 MG
SDP

Catalog No. 5000208259
Encompass_Preferred

1-Ethyl-1H-pyrazole-5-carboxylic acid-d5 is the deuterium-labeled isotopologue of 1-ethyl-1H-pyrazole-5-carboxylic acid intended for use as an internal standard or tracer in analytical workflows. It has CAS number 2735654-39-2, molecular formula C6H3D5N2O2, molecular weight 145.17, and is supplied at high isotopic purity for research use only.

  • Deuterium labeled (d5)
  • Intended as an internal standard or tracer for NMR, GC-MS, and LC-MS
  • High purity (99.9%)
  • Molecular formula C6H3D5N2O2; molecular weight 145.17
  • Available as a 50 MG package
  • For research use only; not validated for medical applications

Catalog No. 50-002-08259 Supplier Medchemexpress LLC Supplier No. HYW077074S50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Ethyl-1H-pyrazole-5-carboxylic acid-d5 is the deuterium-labeled isotopologue of 1-ethyl-1H-pyrazole-5-carboxylic acid intended for use as an internal standard or tracer in analytical workflows. It has CAS number 2735654-39-2, molecular formula C6H3D5N2O2, molecular weight 145.17, and is supplied at high isotopic purity for research use only.

  • Deuterium labeled (d5)
  • Intended as an internal standard or tracer for NMR, GC-MS, and LC-MS
  • High purity (99.9%)
  • Molecular formula C6H3D5N2O2; molecular weight 145.17
  • Available as a 50 MG package
  • For research use only; not validated for medical applications

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.