Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-2,3-dimethylimidazolium chloride, 97%

Catalog No. AAH5922518
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5922518 50 g
AAH5922506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5922518 Supplier Thermo Scientific Chemicals Supplier No. H5922518
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 92507-97-6 | C7H13N2 | 125.19 g/mol

It is used reaction media for chemical processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used reaction media for chemical processes.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92507-97-6
Molecular Formula C7H13N2
Molecular Weight (g/mol) 125.19
MDL Number MFCD03093290
InChI Key IRGDPGYNHSIIJJ-UHFFFAOYSA-N
Synonym 1-ethyl-2,3-dimethylimidazolium chloride, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium chloride, ksc496m4d, 1,2-dimethyl-3-ethylimidazolium chloride, 1-ethyl-2,3-dimethyl-imidazolium chloride, 1-ethyl-2,3-dimethylimidazol-3-ium chloride, 1-ethyl-2,3-dimethylimidazol-1-ium chloride, 3-ethyl-1,2-dimethylimidazol-1-ium chloride, 1-ethyl-2,3-dimethylimidazolium chloride t
PubChem CID 2734165
IUPAC Name 1-ethyl-2,3-dimethylimidazol-3-ium;chloride
SMILES CCN1C=C[N+](C)=C1C

Specifications

Quantity 50 g
Sensitivity Moisture sensitive
Formula Weight 160.64
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium chloride
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.