Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-2-methylpyridinium Bromide 98.0+%, TCI America™
SDP

Catalog No. E12405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E12405G 5 g
E12401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E12405G Supplier TCI America Supplier No. E12405G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32353-50-7
Molecular Formula C8H13BrN+
Molecular Weight (g/mol) 203.103
MDL Number MFCD01208017
InChI Key QTXZDFXPUPKBJC-UHFFFAOYSA-N
PubChem CID 123134705
IUPAC Name 1-ethyl-2-methylpyridin-1-ium;hydrobromide
SMILES CC[N+]1=CC=CC=C1C.Br

Specifications

Melting Point 95°C
Quantity 5 g
Formula Weight 202.10
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-2-methylpyridinium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.