Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-guanylthiourea Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. E04195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04195G Supplier TCI America Supplier No. E04195G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 381670-29-7
Molecular Formula C4H12ClN5S
Molecular Weight (g/mol) 197.685
MDL Number MFCD00191430
InChI Key LLSXLCPLOKTRCV-UHFFFAOYSA-N
Synonym 1-Amidino-3-ethylthiourea Hydrochloride, 1-Carbamimidoyl-3-ethylthiourea Hydrochloride
PubChem CID 60160746
IUPAC Name 1-(diaminomethylideneamino)-3-ethylthiourea;hydrochloride
SMILES CCNC(=S)NN=C(N)N.Cl

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 5 g
Formula Weight 182.67
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Ethyl-3-guanylthiourea Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.