Learn More
Strem, An Ascensus Company CAS# 174899-82-2. 5g. 1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide, 99% [EMIIm]. MFCD03788927
SDP
Catalog No. 5090109023
Encompass
CAS# 174899-82-2. 5g. 1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide, 99% [EMIIm]. MFCD03788927. Molecular Weight: 391.31. Molecular Formula: [C6H11N2]+[N(SO2CF3)2]ae. Color/form: colorless to pale yellow liq. Strem# 07-0579. http://www.strem.com/catalog/v/07-0579/
Catalog No. 50-901-09023 Supplier Strem, An Ascensus Company Supplier No. 0705795G
CAS# 174899-82-2. 5g. 1-Ethyl-3-methylimidazolium bis(trifluoromethylsulfonyl)imide, 99% [EMIIm]. MFCD03788927. Molecular Weight: 391.31. Molecular Formula: [C6H11N2]+[N(SO2CF3)2]ae. Color/form: colorless to pale yellow liq. Strem# 07-0579. http://www.strem.com/catalog/v/07-0579/