Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Ethyl-3-Methylimidazolium Bromide

Catalog No. 894835GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-483-5GF 50 g
89-483-50GF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 89-483-5GF Supplier Sigma Aldrich Supplier No. 894835GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Ethyl-3-methylimidazolium bromide can be used to prepare: 1-ethyl-3-methylimidazolium bis((trifluoromethyl)sulfonyl)imide 1-ethyl-3-methylimidazolium tetrafluoroborate(EMI)[Cd(BTC)] (EMI = 1-ethyl-3-methylimidazolium, BTC = 1,3,5-benzenetricarboxylate), a coordination polymerHydrophobic, highly conductive imidazolium molten salt melting at ambient temp.

Specifications

CAS 258864-54-9
Molecular Formula C6H11BrN2
Linear Formula C6H11BrN2
MDL Number MFCD03427610
Quantity 50 g
Molecular Weight (g/mol) 191.07 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.