Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium chloride, 97%

Catalog No. AC354080050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC354080050 5 g Glass bottle
AC354080250 25 g Glass bottle
AC354081000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC354080050 Supplier Thermo Scientific Chemicals Supplier No. 354080050
  • $49.50
    $39.65 / Each of 1

    Save $9.85

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 65039-09-0 | C6H11ClN2 | 146.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ionic liquid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65039-09-0
Molecular Formula C6H11ClN2
Molecular Weight (g/mol) 146.62
MDL Number MFCD00074843
InChI Key BMQZYMYBQZGEEY-UHFFFAOYSA-M
Synonym 1-ethyl-3-methylimidazolium chloride, 1-ethyl-3-methyl-1h-imidazol-3-ium chloride, emim cl, unii-ph90aq1e93, 1-ethyl-3-methylimidazoliumchloride, 1-methyl-3-ethylimidazolium chloride, 1-ethyl-3-methylimidazol-3-ium chloride, emim-cl, basionics™ st 80, dsstox_cid_21180
PubChem CID 2734160
ChEBI CHEBI:61327
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;chloride
SMILES [Cl-].CCN1C=C[N+](C)=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 77.0°C to 79.0°C
Color Yellow
Flash Point 186°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Viscosity 61.2 mPa.s (80°C)
Formula Weight 146.62
Percent Purity 97%
Physical Form Agglomerating Crystalline Powder
Chemical Name or Material 1-Ethyl-3-methylimidazolium chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.