Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Ethyl-3-Methylimidazolium Iodide

Catalog No. 7130315G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-303-15G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 71-303-15G Supplier Sigma Aldrich Supplier No. 7130315G
Only null left
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Ethyl-3-methylimidazolium iodide is an ionic liquid that can be prepared by reacting methylimidazole with iodoethane. The addition of EMImI to 1-ethyl-3-methylimidazolium tetrafluoroborate (EMImBF4), increase its capacitance while developing electric double-layer capacitors (EDLCs) based on EMImBF4.EMImI reacts with aluminum chloride to form 1-ethyl-3-methylimidazolium halogenoaluminate ionic liquid, [emim]I-(AlCl3)x, which is an excellent solvent for Friedel-Crafts acylation of ferrocene.

Specifications

CAS 56511-17-2
Linear Formula C9H20IN
Quantity 10 g
MDL Number MFCD11846110
Synonym EMIMI
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.