Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Ethyl-3-methylimidazolium methanesulfonate, 99%, Thermo Scientific™

Catalog No. AAH2691309
Change view
Click to view available options
Quantity:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2691309 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2691309 Supplier Thermo Scientific Chemicals Supplier No. H2691309

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as an electrolyte for the electrooxidative polymerization of pyrrole. Used for electroplating of aluminum onto steel, electroplating of chromium,„ electropolishing of steel and other metal surfaces. In polymer and biopolymer processing used as„ solvent for regenerating cellulose,„ enabling technology for manufacturing cellulose, derivatives„ dissolution and separation of lignocellulosic biomas.

Solubility
Soluble in water. Soluble in acetone, methanol .

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 145022-45-3
Flash Point 66°C (152°F)
Molecular Formula C7H14N2O3S
Quantity 10g
MDL Number MFCD06798171
UN Number UN3265
Synonym 1-ethyl-3-methylimidazolium methanesulfonate, 1-ethyl-3-methylimidazolium mesylate, basionics'st 35, basionics™ st 35, acmc-1c0dz, dsstox_cid_29075, dsstox_rid_83294, dsstox_gsid_49219, 1-ethyl-3-methyl imidazolium methyl sulfonate, 1-ethyl-3-methylimidazolium me-thanesulfonate
InChI Key IXLWEDFOKSJYBD-UHFFFAOYSA-M
SMILES CCN1C=C[N+](=C1)C.CS(=O)(=O)[O-]
IUPAC Name 1-ethyl-3-methylimidazol-3-ium;methanesulfonate
Molecular Weight (g/mol) 206.26
PubChem CID 16211849
Formula Weight 206.26
Percent Purity 99%
Chemical Name or Material 1-Ethyl-3-methylimidazolium methanesulfonate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.